Learn More
SI Analytics™ Ostwald Viscometer
Product Code. 10489342 Shop All SI Analytics Products
Change view
Click to view available options
Approximate Constant:
0.01K
0.03K
Size:
0.4mm Diameter
0.5mm Diameter
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Approximate Constant Size Unit Size
10489342 0.03K 0.5mm Diameter Each
10693181 0.01K 0.4mm Diameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10489342 Supplier SI Analytics™ Supplier No. 285404022
  • ONLINE EXCLUSIVE PRICE
    304.81€
    200.00€ / 1 pièces
    Économisez 104.81€
    34% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Ostwald Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high or low temperature measurements
  • With ring marks
  • Without constants
  • For manual measurements
  • Filling quantity of 3mL
  • Overall length of 220mm
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Approximate Constant 0.03K
Includes Ring Mark
For Use With (Application) For Manual Measurements
Size 0.5mm Diameter
Volume (Metric) 3 mL