Learn More
Velp Scientifica™ OXITEST Oxidation Stability Reactor
BIOPHARMA
Product Code. 11743479 Shop All Velp Products
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11743479 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11743479 Supplier Velp Scientifica™ Supplier No. F30900248
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Provides high quality, added value information concerning oxidative stability in foods, oils and fats. Velp Scientifica™ OXITEST Oxidation Stability Reactor is an innovative solution controlled by the powerful OXISoft™.
  • Prediction of the oxidative stability during shelf-life studies
  • Tests the product at defined time intervals; builds an experimental curve
  • Evaluation of the adequacy of storage conditions
  • Evaluation of the best packaging solution
  • Comparison of the oxidative stability of different formulas for food preparations
  • Evaluation of the oxidative stability of vegetable oils of different botanical origin
  • Evaluation of the effectiveness of antioxidants
  • Information on product oxidation when the oxidation flex is not visible
  • Especially useful for products with a low fat content (4-5%)
  • Product oxidation can be achieved by combining the OXITEST with GC
  • Oxidative stability test of incoming raw materials and outgoing finished products
  • Intuitive OXISoft™ software – easy to learn and use
  • Overpressure safety valve
  • Alarms for damaged probe and out-of-range temperature
  • Choice of language for the management software
  • No sample preparation – works on the whole sample (fat included)
  • Twin chambers for duplicate or parallel analysis
  • Manage up to 4 OXITESTs with the same software
  • Indestructible titanium sample holders and chambers
  • Hermetic, sealed closure of the chambers
Specifications
For Use With (Application) Food and Feed, Chemical, Cosmetic, Lubricant Testing
Quantity 1