Learn More
Oxyquinoline Sulfate, TRC
Product Code. 30416847
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416847 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416847 Supplier Toronto Research Chemicals Supplier No. TRCO876990200MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 8-Hydroxyquinoline sulfate
CAS 134-31-6
Molecular Formula 2 C9 H7 N O . H2 O4 S
Quantity 200 mg
Synonym 8-Quinolinol Sulfate (2:1), 8-Quinolinol','sulfate (2:1) (salt) (8CI,9CI), 8-Hydroxyquinoline sulfate, 8-Hydroxyquinoline sulfuric acid salt, 8-Hydroxyquinoline sulphate, 8-Quinolinol sulfate, 8-Quinolinol sulfate (salt), 8-Quinolinol sulphate, Chinosol, Criptonol, Cryptonol, Hydroxyquinoline sulfate, Khinozol, Octofen, Oxine sulfate, Oxyquinol, Oxyquinoline sulfate, Quinolin-8-ol sulfate (2:1), Quinosol, Quinosol extra, Solvochin-Extra, Sunoxol, Superol
InChI Formula InChI=1S/2C9H7NO.H2O4S/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;1-5(2,3)4/h2*1-6,11H;(H2,1,2,3,4)
SMILES Oc1cccc2cccnc12.Oc3cccc4cccnc34.OS(=O)(=O)O
IUPAC Name quinolin-8-ol;sulfuric acid
Molecular Weight (g/mol) 388.39
Formula Weight 388.0729