Learn More
Palonosetron-3-ene Hydrochloride, TRC
Product Code. 30136034
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136034 250 mg 250mg
30008577 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136034 Supplier Toronto Research Chemicals Supplier No. TRCP165825250MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 135729-55-4
Molecular Formula C19 H22 N2 O . Cl H
Molecular Weight (g/mol) 330.85
Synonym 1H-Benz[de]isoquinolin-1-one, 2-(3S)-1-azabicyclo[2.2.2]oct-3-yl-2,4,5,6-tetrahydro-, hydrochloride (1:1), 1H-Benz[de]isoquinolin-1-one, 2-(1-azabicyclo[2.2.2]oct-3-yl)-2,4,5,6-tetrahydro-, monohydrochloride, (S)-, 1H-Benz[de]isoquinolin-1-one, 2-(3S)-1-azabicyclo[2.2.2]oct-3-yl-2,4,5,6-tetrahydro-, monohydrochloride (9CI), 2-[(3S)-1-Azabicyclo[2.2.2]oct-3-yl] 2,4,5,6-tetrahydro-1H-benzo[de]isoquinolin-1-one hydrochloride, RS 25259-007, RS 42358-197
IUPAC Name 2-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-5,6-dihydro-4H-benzo[de]isoquinolin-1-one;hydrochloride
SMILES Cl.O=C1N(C=C2CCCc3cccc1c23)[C@@H]4CN5CCC4CC5
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C19H22N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,11,13,17H,1,3,5,7-10,12H2;1H/t17-;/m1./s1
Formula Weight 330.1499
Percent Purity >95%
Chemical Name or Material Palonosetron-3-ene Hydrochloride
Grade HPLC