Learn More
Gilac™ Patient Drinking Cup with Nose Cut-out, Green
Product Code. 17958991
Change view
Click to view available options
Capacity (Metric):
0.25 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17958991 0.25 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17958991 Supplier Gilac™ Supplier No. G120508
  • ONLINE EXCLUSIVE PRICE
    10.56€
    7.06€ / 1 pièces
    Économisez 3.50€
    33% de réduction
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Patient Drinking Cup with Nose Cut-out allows drinking without putting the head back.
  • Pack of 10 patient drinking cup with nose cut-out.
  • Drinking cup with graduations (in mL).
  • Allows drinking more easily.
Specifications
Material Polypropylene
Capacity (Metric) 0.25 L
Capacity (English) 8.5 oz.
Quantity 10 Pk.
Product Type Tumbler Cup