Learn More
Cytiva PEEK Tubing
Product Code. 11555724 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.150 mm
0.5 mm
0.75 mm
1 mm
Unit Size:
2m
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
11555724 0.5 mm 2m
11360012 0.150 mm Each
10328439 1 mm Each
11330012 0.75 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11555724 Supplier Cytiva Supplier No. 18111368
En stock
Add to basket
Edge
Add to basket
PEEK tubing comes in a variety of internal diameter sizes. Cytiva Lifescience™ PEEK Tubing is an important chromatography accessory.
  • Polyetheretherketone (PEEK)
  • A variety of sizes are available
TRUSTED_SUSTAINABILITY
Specifications
Type Tubing
Color Orange
Description PEEK Tubing, 2 m, i.d. 0.5 mm, o.d. 1/16 in.
Diameter (Metric) Inner 0.5 mm
Diameter (Metric) Outer 1.58 mm
Overall Length (Metric) 2 m
Material PEEK