Learn More
3M™ PELTOR™ Hygiene Kit
Product Code. 15710108 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Bull's Eye III Earmuffs
Bull's Eye™ I Earmuffs
Bull's Eye™ II Earmuffs
H6 Series Earmuffs
Optime Push To Listen Earmuffs
Optime™ I Earmuffs
Optime™ II Earmuffs
Optime™ III Earmuffs
Worktunes Headset
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15710108 Worktunes Headset Each
15739967 Bull's Eye™ I Earmuffs Each
15749967 Bull's Eye™ II Earmuffs Each
15719957 Bull's Eye III Earmuffs Each
15729957 H6 Series Earmuffs Each
15779707 Optime™ I Earmuffs Each
15709647 Optime™ II Earmuffs Each
15709957 Optime™ III Earmuffs Each
15719927 Optime Push To Listen Earmuffs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15710108 Supplier 3M™ Supplier No. 7100101874
  • ONLINE EXCLUSIVE PRICE
    27.00€
    13.75€ / 1 pièces
    Économisez 13.25€
    49% de réduction
Expédition estimée: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
3M™ PELTOR™ Hygiene Kit contains replacement sealing rings and muffler pads which can be replaced in a few easy steps.
  • Kit contains replacement sealing rings and muffler pads which can be replaced in a few easy steps
  • Extends the life of earmuffs
  • Ensures good function and hygiene
  • Avoid wearing uncomfortable earmuffs
  • Pack contains 2 x foam inserts and 2 sealing rings
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hygiene Kit
For Use With (Equipment) Worktunes Headset
Includes Foam Inserts and Sealing Rings