Learn More
3M™ PELTOR™ Optime™ III Earmuffs
Product Code. 15789947
Change view
Click to view available options
Model:
H540A-461-GB
H540B-474-GB
H540P3AF-413-SV
H540P3B-413-SV
H540P3E-413-SV
H540P3G-413-SV
H540P3H-413-SV
H540P3K-413-SV
Unit Size:
Pack of 10
Pack of 20
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15789947 H540P3B-413-SV Pack of 20
15739017 H540P3E-413-SV Pack of 20
15709737 H540P3AF-413-SV Pack of 6
15709927 H540B-474-GB Pack of 20
15759697 H540A-461-GB Pack of 10
15789917 H540P3H-413-SV Pack of 20
15799917 H540P3K-413-SV Pack of 20
15799947 H540P3G-413-SV Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15789947 Supplier 3M™ Supplier No. 7000107981
This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products
This item is not returnable. View return policy
3M™ PELTOR™ Optime™ III Earmuffs are a high performance hearing protector, developed for use in extremely noise environments like airports, marine engine rooms, power stations and print works.
  • Ideal hearing protection against noise arising from extremely noisy environments like airports, marine engine rooms, power stations and print works
  • Double casings technology helps minimize resonance effect
  • Large space inside cup helps reduce moisture and heat build-up
  • Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
  • Easy to replace cushions and inserts helps keep them hygienically clean
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Red
Certifications/Compliance CE Approved, EN 352-3: 2002
Model H540P3B-413-SV
Noise Reduction Rating (NRR) 34 dB
Product Type Ear Muffs
Dielectric No
Style Helmet-mounted