Learn More
3M™ PELTOR™ WorkTunes™ Pro AM/FM Radio Headset
Product Code. 15750098 Shop All 3M Products
Change view
Click to view available options
Noise Level:
31 dB
32 dB
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Noise Level Unit Size
15750098 31 dB Pack of 10
15740098 32 dB Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15750098 Supplier 3M™ Supplier No. 7100089440
  • ONLINE EXCLUSIVE PRICE
    1143.05€
    803.42€ / 10 pièces
    Économisez 339.63€
    29% de réduction
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
3M™ PELTOR™ WorkTunes™ Pro AM/FM Radio Headset helps protect you from potentially harmful workplace noise while at the same time enjoy listening to the built in AM/FM radio.
  • Protect your hearing and listen to AM/FM radio at the same time with built-in antenna
  • Digital station search and voice guided menu system with announced frequencies
  • 3.5 mm listen only stereo input (limited to 82 dB) for connection to external devices (e.g. mobile phone, radio, two-way radio and iPod)
  • Auto power off turns headset off after 4 hours of non-use, to save energy and a low-battery warning informs you when to change batteries
  • To reduce the corrosion caused by sweat the electronics are located in the outer part of the cup
  • Store up to 5 stations on the headset
  • Available in both headband and helmet attached versions (black)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Radio Headset
Model HRXS221P3E
Noise Level 31 dB