Learn More
Pentafluorophenyl methacrylate, 98%, BLD Pharm™
Product Code. 31089548
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31089548 1 g
31076927 25 g
31089539 5 g
31079584 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31089548 Supplier BLD Pharm Supplier No. BD1470381g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
(Stabilized with MEHQ)
Specifications
CAS 13642-97-2
Molecular Formula C10H5F5O2
MDL Number MFCD00042332
Quantity 1 g
SMILES CC(=C)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 252.14
Percent Purity 98%
Chemical Name or Material Pentafluorophenyl methacrylate