Learn More
Perfluorophenyl acrylate, 98%, BLD Pharm™
Product Code. 31428169
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31428169 250 mg
31421544 100 g
31424691 5 g
31418507 25 g
31420562 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31428169 Supplier BLD Pharm Supplier No. BD147035250mg
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Stabilized with 500ppm MEHQ
Specifications
CAS 71195-85-2
Molecular Formula C9H3F5O2
MDL Number MFCD00042330
Quantity 250 mg
SMILES C=CC(OC1=C(F)C(F)=C(F)C(F)=C1F)=O
Molecular Weight (g/mol) 238.11
Percent Purity 98%
Chemical Name or Material Perfluorophenyl acrylate