Learn More
Liebherr™ Performance Underbench STATIC Lab Freezer
Product Code. 30464307 Shop All Liebherr Appliances Products
Change view
Click to view available options
Drawers:
2 white freezer drawers
Not included
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Drawers Unit Size
30464307 Not included Each
30464304 2 white freezer drawers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 30464307 Supplier Liebherr™ Supplier No. SFTSG1501
  • ONLINE EXCLUSIVE PRICE
    2900.00€
    2864.53€ / 1 pièces
    Économisez 35.47€
    1% de réduction
 Restricted Item
Expédition estimée: 06-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Secure, accurate and ultra energy-efficient freezer storage, the Liebherr™ Underbench Lab Freezer is designed for general purpose applications with Liebherr build-quality you can rely on.
  • Underbench lab freezer, width 600 mm, gross capacity 101 L
  • Temperature range -9°C to -30°C ± 5°C temperature stability tested to EN60068-3
  • SmartFrost static refrigeration, manual defrost
  • white exterior, commercial-grade moulded polystyrol interior
  • Refrigerant R 290, connection rating 2.0 A, 220-240V/50/1-ph, energy consumption 180 kWh/a
  • 69 L net capacity
  • 2 white-coated wire shelves
  • Multifunctional digital controller with high resolution display and connectivity options
  • Door lock, reversible hinges, antimicrobial lever handle
  • 45 kg shelf load
  • 5-year extended warranty option available in select countries
Specifications
Type Lab freezer
Capacity (English) 26.7 gal.
Capacity (Metric) 101 L
Temperature Range -9°C to -30°C
Temperature Resolution Display 0.1 °C
Temperature Sensor NTC
Temperature Stability ±5 K
Door Style Reversible, Self-closing, Fitted Lock
No. of Doors 1
Door Material White Powder-coated steel
Cabinet Material Scratch-resistant Painted Steel
Chamber Material Polystyrene
Interior Commercial-grade moulded polystyrol
Shelves 2
Drawers Not included
Certifications/Compliance ISO 9001, ISO 14001, CE, DIN 13277
Style Underbench
Display Monochrome display in door
Color White
Condenser Type Low-maintenance rear-wall wire-on-tube
Control Type Touch control / digital display in door
For Use With (Application) General-purpose lab freezer storage
Ports 1 x 10 mm
Product Line Scientific Performance
Warranty 2 Years Parts and Labor, 5 Year Optional Extended Warranty
Defrost Manual
Model SFTsg 1501
Monitoring Options SmartCoolingHub
Connectivity LAN/WiFi module optional
Amperage 2.0 A
Plug Type CEE 7/7
Voltage 220/240 V
Frequency 50 Hz
Phase 1, single phase
Wattage 110 W
Energy Usage 180 kWh/a
Green Features Low energy consumption
Compressor CFC-free
Refrigerant R 600a
Shipping Weight (Metric) 37 kg