Learn More
Thermo Scientific™ Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Shop All Thermo Scientific ProductsDescription
Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems; all designed for durability and fitness for purpose.
Select High Quality Consumables for Improved Performance
Choose from the following Peristaltic Pump Tubing:
- Peristaltic Pump Tubing - dia.0,508 -Orange/Yellow -PVC
- Peristaltic Pump Tubing - dia.0,381 -Orange/Green - PVC
- Peristaltic Pump Tubing - dia.0,762 - Black/Black -PVC
- Peristaltic Pump Tubing - dia.0,254 - Orange/Blue -PVC
- Peristaltic Pump Tubing - dia.1,295 - Gray/Gray -Santoprene
- Peristaltic Pump Tubing - dia.3,175 - Black/White -Santoprene
Specifications
Specifications
| Type | Peristaltic Pump Tubing |
| For Use With (Equipment) | iCAP MX and iCAP Q/Qnova Series |
| Material | PVC |
| Diameter (Metric) | 0.762 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?