Learn More
PF-AKT400, MedChemExpress
Product Code. 16476807
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476807 5 mg 5mg
16455076 100 mg 100mg
16434108 10 mg 10mg
16410467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476807 Supplier Medchem Express Supplier No. HY10721
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress PF-AKT400 is a broadly selective, potent, ATP-competitive Akt inhibitor, displays 900-fold greater selectivity for PKBα (IC50=0.5 nM) than PKA (IC50=450 nM).
PF-AKT400 is a broadly selective, potent, ATP-competitive Akt inhibitor, displays 900-fold greater selectivity for PKBα (IC50=0.5 nM) than PKA (IC50=450 nM).
Specifications
Chemical Name or Material PF-AKT400
CAS 1004990-28-6
Color White
Purity Grade Notes Research
Molecular Formula C20H22F2N6O
Quantity 5 mg
Synonym AKT protein kinase inhibitor
Solubility Information DMSO : ≥ 100 mg/mL (249.73 mM)
SMILES N[C@@](CC1)(CNC(C2=CC=C(F)C=C2F)=O)CN1C3=C4C(NC=C4CC)=NC=N3
Molecular Weight (g/mol) 400.43
Formula Weight 400.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease