Learn More
WTW™ pH Combination Electrode with Armoring
Product Code. 10263021
Change view
Click to view available options
pH Range:
2 to 12
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10263021 2 to 12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10263021 Supplier WTW™ Supplier No. 109115
  • ONLINE EXCLUSIVE PRICE
    415.00€
    328.49€ / 1 pièces
    Économisez 86.51€
    20% de réduction
Expédition estimée: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Connector Types Watertight plug head connector
Type pH Combination Electrode with Armoring
Filling Solution Type Gel Polymer solid electrolyte (AgCl-free)
Includes Solid electrolyte
Junction Hole junction
pH Range 2 to 12
Temperature (Metric) Operating 0°C to 60°C