Learn More
Plum™ pH Neutral Eyewash Bottle
Product Code. 10511593
Change view
Click to view available options
Capacity (Metric):
200 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10511593 200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10511593 Supplier Plum™ Supplier No. 4752
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
pH Neutral will neutralize acid and base substances and provide effective protection against serious eye injuries
  • The product features phosphate buffer properties with documented efficacy in chemicals-based eye injuries
  • All standard pH Neutral products (except DUO) are supplied with dust cap
  • Recommend wash time: 2 min.
  • 4.9 % sterile phosphate buffer for neutralisation of acid and base that come into contact with the eye
  • Washing with pH Neutral is recommended to be followed by washing with Plum Eyewash
  • Easily manageable bottles
  • Ergonomic eye-cups for effective flow
  • DUO eye-cup for simultaneous washing of both eyes
  • User-friendly design allows the bottles to be quickly opened with a single hand
  • Disposable bottles; do not reuse
  • CE-labelled bottles, products with 3-year expiry date
Specifications
Capacity (Metric) 200 mL
Includes 10 x 200 mL Bottles of pH Neutral