Learn More
Magle Life Sciences™ Phadebas™ Saliva Amylase-detection Forensic Paper
Product Code. 12895552 Shop All Magle Life Sciences Products
Change view
Click to view available options
Dimensions (L x W):
300 x 195 mm
395mm x 15m roll
Unit Size:
Each
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12895552 300 x 195 mm Pack of 12
12825562 395mm x 15m roll Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12895552 Supplier Magle Life Sciences™ Supplier No. 1314
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Used for presumptive testing of suspicious stains. Phadebas™ Saliva Amylase-detection Forensic Paper is based on the Phadebas Amylase Test tablets but follows a separate method developed by the Forensic Biology labs.
Pre-manufactured papers;
  • Are applicable directly from package;
  • Include Batch number and Expiry date;
  • Are easy to handle and use;
  • Have negligible batch variations;
  • Give no difference between labs and consequently have higher credibility as evidence material
Specifications
Dimensions (L x W) 300 x 195 mm