Learn More
Phenazine, 97%, BLD Pharm™
Product Code. 31336429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
1 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31336429 500 g
31319547 10 g
31330253 100 g
31318280 1 g
31329558 5 g
31334331 25 g
31335656 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31336429 Supplier BLD Pharm Supplier No. BD14396500g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 92-82-0
Molecular Formula C12H8N2
MDL Number MFCD00005023
Quantity 500 g
SMILES C12=NC3=CC=CC=C3N=C1C=CC=C2
Molecular Weight (g/mol) 180.21
Percent Purity 97%
Chemical Name or Material Phenazine