missing translation for 'onlineSavingsMsg'
Learn More

Phenylglyoxylic acid, 98%

Product Code. 11300154
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11300154 25 g 25g
11390144 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11300154 Supplier Thermo Scientific Alfa Aesar Supplier No. L00648.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-73-4 | C8H6O3 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-73-4
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00002575
InChI Key FAQJJMHZNSSFSM-UHFFFAOYSA-N
Synonym benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid
PubChem CID 11915
ChEBI CHEBI:18280
IUPAC Name 2-oxo-2-phenylacetic acid
SMILES OC(=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 62°C to 67°C
Density 1.38
Boiling Point 147°C to 151°C (12 mmHg)
Quantity 25 g
Beilstein 606718
Formula Weight 150.13
Percent Purity 98%
Chemical Name or Material Phenylglyoxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.