Learn More
Abcam Phosphoglucomutase Activity Assay Kit (Fluorometric)
Product Code. 32112903 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32112903 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32112903 Supplier Abcam Supplier No. ab211104100Test
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Measure PGM1 activity in Cell/Tissue Extracts with Phosphoglucomutase Activity Assay Kit AB211104. Sensitivity ≤ 1 μU. Cited in 1 publication. Use with Microplate reader.
Measure PGM1 activity in Cell/Tissue Extracts with Phosphoglucomutase Activity Assay Kit AB211104. Sensitivity ≤ 1 μU. Cited in 1 publication. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests
Regulatory Status RUO