Learn More
Phosphorus lump, Red, Puratronic™, 99.999+% (metals basis)
Product Code. 11340899
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340899 10 g 10g
11320899 1 g 1g
11330899 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 11340899 Supplier Thermo Scientific Alfa Aesar Supplier No. 010670.09
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
CAS: 7723-14-0 | P | 30.97 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7723-14-0
Molecular Formula P
Molecular Weight (g/mol) 30.97
MDL Number MFCD00133771
InChI Key OAICVXFJPJFONN-UHFFFAOYSA-N
Synonym red, acmc-20 ajw2
PubChem CID 5462309
ChEBI CHEBI:30207
IUPAC Name phosphorus
SMILES [P]
Specifications
Color Brown to Red
Physical Form Liquid
Quantity 10 g
Ignition Point 300°C
Appearance Powder or Lump
Vapor Pressure 7100 hPa (5325 mm Hg)
Assay Percent Range Puratronic™, 99.999+% (metals basis)
Density 1.701 g/cm3 (14.195 lb./gal)
UN Number 1338
Solubility Information Miscible with water
Chemical Name or Material Phosphorus lump, red