Learn More
Sartorius Pipette Calibration Draft Shield Cubis II UHR
Product Code. 31662883 Shop All Sartorius Products
Change view
Click to view available options
Product Type:
Pipette Calibration Draft Shield
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31662883 Pipette Calibration Draft Shield Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 31662883 Supplier Sartorius Supplier No. YCP01MPS
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
  • The Cubis™ II UHR Pipette Calibration Accessory provides an ideal solution to transform a Cubis™ II UHR balance into a pipette calibration system.
  • This way, the balance offers maximum versatility for end users who use UHR balances for weighing applications and also want to use them to calibrate or check their pipettes.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pipette Calibration Draft Shield