Learn More
Pirimicarb-d6 (dimethyl-d6-carbamoyl), CDN
Product Code. 30319692
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319692 0.01 g 0.01g
30320315 0.005 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319692 Supplier CDN Supplier No. CDND78220.01G
 Restricted Item
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pirimicarb D6 (dimethylcarbamate D6)
CAS 1015854-66-6
Molecular Formula C11 2H6 H12 N4 O2
Quantity 0.01 g
InChI Formula InChI=1 S/C11H18N4O2/c1-7-8(2)12-10(14(3)4)13-9(7)17-11(16)15(5)6/h1-6H3/i5D3,6D3
SMILES [2 H]C([2 H])([2 H])N(C(=O)Oc1nc(nc(C)c1C)N(C)C)C([2 H])([2 H])[2 H]
IUPAC Name [2-(dimethylamino)-5,6-dimethylpyrimidin-4-yl] N,N-bis(trideuteriomethyl)carbamate
Molecular Weight (g/mol) 244.32
Formula Weight 244.1806 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity