Learn More
Abcam Plasmin Activity Assay Kit (Fluorometric)
Product Code. 32112047 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32112047 1 x 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32112047 Supplier Abcam Supplier No. ab3246301x96Tests
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Measure Plasminogen activity in 1h in Biofluids with Plasmin Activity Assay Kit AB324630. Sensitivity = 8.66 μU. Use with Microplate (12 x 8 well strips).
Measure Plasminogen activity in 1h in Biofluids with Plasmin Activity Assay Kit AB324630. Sensitivity = 8.66 μU. Use with Microplate (12 x 8 well strips).
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 96 Tests
Regulatory Status RUO