Learn More
Gilac™ Plates
Artikelnummer. 17978541
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
10 Pk.
Pakningsstørrelse:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
17978541 10 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17978541 Leverandør Gilac™ Leverandørnr. G332621
  • ONLINE EXCLUSIVE PRICE
    32.55€
    21.80€ / 1 pièces
    Économisez 10.75€
    33% de réduction
Expédition estimée: 09-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Reusable plastic plates with outstanding durability, even in intensive cycle use.
  • Reusable plastic plate.
  • Outstanding durability even in intensive cycle use.
  • Ideal for institutions.
  • Pack of 10.
Tekniske data
Product Type Plate
Material Polypropylene
Color White
Diameter (Metric) 217 mm
Height (English) 1.4 in.
Height (Metric) 35 mm
Quantity 10 Pk.