Learn More
Platinum(II) chloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 10025-65-7 |
| Quantity | 100 mg |
| Molecular Formula | Cl2Pt |
| MDL Number | MFCD00011181 |
| SMILES | [Pt+2].[Cl-].[Cl-] |
| Molecular Weight (g/mol) | 265.98 |
| Percent Purity | 98% |
| Chemical Name or Material | Platinum(II) chloride |