Learn More
Testo™ Pocket Sized RPM Meter
Product Code. 10324560
Change view
Click to view available options
Range:
0°C to 50°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10324560 0°C to 50°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 10324560 Supplier Testo™ Supplier No. 0560 0460
  • ONLINE EXCLUSIVE PRICE
    206.05€
    167.00€ / 1 pièces
    Économisez 39.05€
    18% de réduction
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Specifications
Product Type RPM Meter
Includes Reflective markers, protective cap, hand strap, belt bag
Range 0°C to 50°C
For Use With (Application) Non-contact rpm measurements