Learn More
Euro Protection™ Polyamide Gloves with Polyurethane Coating
Click to view available options
Size:
6
7
8
9
10
11
Color:
Black
Gray
White
Unit Size:
10 pairs
Description
- 1 per pack
- CE Mark certification
- Polyamide construction
- Polyurethane coating in various colors
Specifications
Specifications
| Certifications/Compliance | CE Mark |
| Coated Area | Palm |
| Description | Polyamide and Copper Wire |
| Coating Material | Polyurethane |
| Color | White |
| Material | Polyamide, Copper |
| Product Type | Gloves |
| Size | 7 |
| Glove Cuff Style | Elasticated |
| Packaging Type | Individual |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction