Learn More
Masterflex™ Polycarbonate Flowmeters with Valve
Product Code. 15233767 Shop All Masterflex Products
Change view
Click to view available options
Flow Rate Range:
0.1 to 1.2 LPM
4 to 50 LPM
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Range Unit Size
15233767 0.1 to 1.2 LPM Each
15223767 4 to 50 LPM Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15233767 Supplier Masterflex™ Supplier No. 3290044
  • ONLINE EXCLUSIVE PRICE
    184.00€
    126.84€ / 1 pièces
    Économisez 57.16€
    31% de réduction
View all promotions
Expédition estimée: 08-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type Variable Area Flowmeter
Accuracy ±4% (Full Scale)
Float Material Stainless Steel
Pressure Rating 100 psig
Flow Rate Range 0.1 to 1.2 LPM
For Use With (Application) Precision-adjusting valve for accurate, leak-free flow control
Repeatability ±1%
Temperature (Metric) 65°C