Learn More
Brady™ Polypropylene: ISO Safety Sign - General warning sign
Product Code. 15624677
Click to view available options
Width (Metric):
100 mm
315 mm
Height (Metric):
87 mm
273 mm
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 15624677
Brand: Brady™ 826621
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Sign compliant to ISO 7010.
  • Polypropylene
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC W001-Tri 315-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 273 mm
Thickness 1.501 mm
Width (Metric) 315 mm
Temperature Range (Metric) -40°C to 120°C
Shape Triangular
Languages Global
Color Black on Yellow