Learn More
Thermo Scientific™ POROS™ GoPure™ AAV9 Screening Plate, 96-well, 20 μL
Product Code. 30131540 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30131540 1 Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30131540 Supplier Thermo Scientific™ Supplier No. A65810
  • ONLINE EXCLUSIVE PRICE
    1015.00€
    974.00€ / 1 pièces
    Économisez 41.00€
    4% de réduction
View all promotions
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Thermo Scientific™ POROS™ GoPure™ AAV9 96-well screening plates contain POROS CaptureSelect AAV9 Affinity Resin and provide a versatile platform for sample preparation, handling, and analysis.
POROS GoPure AAV9 96-well screening plates contain POROS CaptureSelect AAV9 Affinity Resin and provide a versatile platform for sample preparation, handling, and analysis. They accommodate multiple samples simultaneously to achieve high throughput and data generation.
These pre-filled screening plates have the following features:
  • High-throughput screening capability enabling multiple conditions to be evaluated in a single plate experiment
  • 20 μL per well, helping preserve sample while maximizing data generation
  • Ready-to-use 96-well format for efficient evaluation of Thermo Scientific resins
Specifications
Concentration 220 μL Resin Slurry/Well
Content And Storage Store at 2°C to 8°C. Do not freeze.
Shipping Condition Wet Ice
Particle Size 50 μm
Form Resin Slurry
Product Line POROS
Quantity 1 Plate
For Use With (Application) Chromatography
Type Affinity Screening Plate
Sample Volume 20 μL
Ligand Type CaptureSelect AAV9 Affinity Ligand
Format 96-well Filter Plate
Capacity (Metric) 800 μL
Show More Show Less
For Research Use Only.