Learn More
Fisherbrand™ Portable Balance
Product Code. 15949345 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity:
220 g
620 g
2200 g
5200 g
Model:
FCS2200
FCS221
FCS5200
FCS621
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Model Unit Size
15949345 620 g FCS621 Each
15939345 220 g FCS221 Each
15909355 2200 g FCS2200 Each
15919355 5200 g FCS5200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15949345
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
En stock
Add to basket
Edge
Add to basket
Regulatory: This product is under eco-contribution tax.
Portable Balance with a slim, stackable design provides basic weighing functionality and competitive performance.
  • Intuitive two-button design makes for easy operation
  • Slim, space-saving design allows for stackable storage
  • Enlarged weighing platform can adequately handle large samples and a variety of objects
TRUSTED_SUSTAINABILITY
Specifications
Capacity 620 g
Display Type LCD
Readability 0.100000 g
For Use With (Application) Weighing
Includes 3 AA (LR6) Batteries
Model FCS621
Stabilization Time 2.000 sec.
Temperature Range (English) 64°F to 77°F
Temperature Range (Metric) 18°C to 25°C
Warranty 2 Years
Weighing Modes Gram, Pound, Ounce, Ounce Troy, Pennyweight
Dimensions (D x W x H) 205 x 140 x 41 mm (8.1 x 5.5 x 1.6 in.)
Width (English) Pan 5 in.
Width (Metric) Pan 125 mm
Depth (English) Pan 5.2 in.
Depth (Metric) Pan 132 mm
Weight (English) 0.9 lb.
Weight (Metric) 0.4 kg