missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3-bromophenyltrifluoroborate, 97%

Product Code. 11304855
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304855 5 g 5g
11394845 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304855 Supplier Thermo Scientific Alfa Aesar Supplier No. L17966.06
 Restricted Item
Expédition estimée: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 374564-34-8 | C6H4BBrF3K | 262.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374564-34-8
Molecular Formula C6H4BBrF3K
Molecular Weight (g/mol) 262.91
MDL Number MFCD02093977
InChI Key NIDYRVJZTADCGO-UHFFFAOYSA-N
Synonym potassium 3-bromophenyl trifluoroborate, potassium 3-bromophenyltrifluoroborate, potassium 3-bromophenyl trifluoroboranuide, amtb081, potassium 3-bromophenyl-trifluoroboranuide, potassium ion 3-bromophenyl trifluoroboranuide, potassium 3-bromophenyl trifluoro borate 1-, potassium 3-bromophenyl-tris fluoranyl boranuide
PubChem CID 23677642
IUPAC Name potassium;(3-bromophenyl)-trifluoroboranuide
SMILES [K+].F[B-](F)(F)C1=CC(Br)=CC=C1

Specifications

Melting Point 178°C to 182°C
Quantity 5 g
Formula Weight 262.91
Percent Purity 97%
Chemical Name or Material Potassium 3-bromophenyltrifluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.