Learn More
Potassium Peroxymonosulfate Sulfate, TRC
Code produit 30138769
Modifier l'affichage
Click to view available options
Quantity:
10 g
100 g
Conditionnement:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
30138769 100 g 100g
30006483 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30138769 Fournisseur Toronto Research Chemicals Code fournisseur TRCP698200100G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 37222-66-5
Molecular Formula H3K5O18S4
Molecular Weight (g/mol) 614.76
Synonym Potassium Peroxymonosulfate Sulfate (K5(HSO5)2(HSO4)(SO4)), Potassium Peroxymonosulfate Sulfate (K5[HSO3(O2)]2(HSO4)(SO4)), Basolan 2448, Caro Salt, Caroat, Caroate, Caro's Salt, Caro's Triple Salt, Curox, L-Gel, Oxone, PS 16, PS 16 (salt), Potassium Peroxymonosulfate Sulfate (2KHSO5.KHSO4.K2SO4), Potassium persulfate triple salt, Urutora San
IUPAC Name pentapotassium;hydrogen sulfate;hydroxy sulfate;sulfate
SMILES [K+].[K+].[K+].[K+].[K+].OOS(=O)(=O)[O-].OOS(=O)(=O)[O-].OS(=O)(=O)[O-].[O-]S(=O)(=O)[O-]
Spécification
Quantity 100 g
InChI Formula InChI=1S/5K.2H2O5S.2H2O4S/c;;;;;2*1-5-6(2,3)4;2*1-5(2,3)4/h;;;;;2*1H,(H,2,3,4);2*(H2,1,2,3,4)/q5*+1;;;;/p-5
Formula Weight 613.6388
Chemical Name or Material Potassium Peroxymonosulfate Sulfate