Learn More
Robin™ Power Mixers
Product Code. 11979607
Change view
Click to view available options
Speed Range:
12 t/min.
2 to 25 t/min.
2 to 40 t/min.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
11979607 2 to 25 t/min. Each
11939667 12 t/min. Each
11969607 2 to 40 t/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11979607 Supplier Robin™ Supplier No. PM033
Expédition estimée: 23-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Specifications
Speed Range 2 to 25 t/min.
Holds 10 L
Product Type Power Mixer
Depth (Metric) 50 cm
Height (Metric) 60 cm
Length (Metric) 56 cm
Weight (Metric) 18.5 kg