Learn More
Dostmann Electronic™ Power Supply
Product Code. 11854392
Change view
Click to view available options
Voltage:
9 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
11854392 9 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11854392 Supplier Dostmann Electronic™ Supplier No. 56000702
  • ONLINE EXCLUSIVE PRICE
    51.95€
    28.00€ / 1 pièces
    Économisez 23.95€
    46% de réduction
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Specifications
Product Type Power Supply
For Use With (Equipment) P700 1-Kanal Universal-Thermometer, P705 2-Kanal Universal-Thermometer, P710 1-Kanal Universal-Thermometer, P715 2-Kanal Universal-Thermometer, P750 Temperature-humidity instrument, P755 multi-function instrument for temperature-humidity-airflow-pressure,
Voltage 9 V
Warranty 15 Months