Learn More
Fisherbrand™ Power Supply for Fisherbrand™ accumet™ AB and XL Benchtop Meters
Product Code. 12850653 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12850653 AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12850653
  • ONLINE EXCLUSIVE PRICE
    69.10€
    45.08€ / 1 pièces
    Économisez 24.02€
    34% de réduction
View all promotions
 Restricted Item
Expédition estimée: 19-01-2027
Add to basket
Add to basket
Universal replacement power supply
  • For use with accumet benchtop meters models: AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 meters
TRUSTED_SUSTAINABILITY
Specifications
Type Power Supply
Electrical Requirements 110/230 VAC, 9 VDC, 18 W, center positive
For Use With (Equipment) AB150, AB200, AB250, XL150, XL200, XL250, XL500, XL600 Meters