Learn More
Honeywell™ POWERCOAT™ Nitri-Knit Gloves
Description
- Supported nitrile glove
- Cotton interlock liner
- Textured finish
- Interlock knit liner gives gloves added strength for resistance to cuts, snags, abrasions and punctures while adding comfort to wearer
- Hand portion is treated with a permanent anti-bacterial and anti-mildew agent
- Unique textured finish provides excellent grip on oily, wet or dry objects
- High nitrile content offers superior resistance to cuts, snags, abrasions and punctures
- Outperforms supported natural rubber, neoprene and vinyl gloves when exposed to solvents, caustics and animal fats
- True dip coated glove has a permanent bond of nitrile to inner-liner, won't peel apart like glued-in liners
- Gloves are free of latex proteins which can cause allergic reactions
Specifications
Specifications
| Certifications/Compliance | EC PPE Category 3, EC Certificate/Attestation Number BP60077212 0001 |
| Coating Material | Nitrile |
| Color | Blue |
| Exterior Finish | Fully Textured |
| Size | 8, Medium |
| Thickness (Metric) | 1 mm |
| Liner Material | Cotton |
| Length (Metric) | 30 cm |