Learn More
Pranlukast, TRC
Product Code. 30424450
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424450 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424450 Supplier Toronto Research Chemicals Supplier No. TRCP701100100MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pranlukast
CAS 103177-37-3
Molecular Formula C27 H23 N5 O4
Quantity 100 mg
Synonym Benzamide','N-[4-oxo-2-(2H-tetrazol-5-yl)-4H-1-benzopyran-8-yl]-4-(4-phenylbutoxy)-, Benzamide','N-[4-oxo-2-(1H-tetrazol-5-yl)-4H-1-benzopyran-8-yl]-4-(4-phenylbutoxy)- (9CI), N-[4-Oxo-2-(2H-tetrazol-5-yl)-4H-1-benzopyran-8-yl]-4-(4-phenylbutoxy)benzamide, 4-Oxo-8-[4-(4-phenylbutoxy)benzoylamino]-2-(tetrazol-5-yl)-4H-1-benzopyran, 8-[4-(4-Phenylbutoxy)benzoyl]amino-2-(5-tetrazolyl)-4-oxo-4H-1-benzopyran, Azlaire, CCN 00401, ONO 1078, ONO-RS 411, Onon, Pranlukast, RS 411, SB 205312
InChI Formula InChI=1S/C27H23N5O4/c33-23-17-24(26-29-31-32-30-26)36-25-21(23)10-6-11-22(25)28-27(34)19-12-14-20(15-13-19)35-16-5-4-9-18-7-2-1-3-8-18/h1-3,6-8,10-15,17H,4-5,9,16H2,(H,28,34)(H,29,30,31,32)
SMILES O=C(Nc1cccc2C(=O)C=C(Oc12)c3nnn[nH]3)c4ccc(OCCCCc5ccccc5)cc4
IUPAC Name N-[4-oxo-2-(1H-tetrazol-5-yl)chromen-8-yl]-4-(4-phenylbutoxy)benzamide
Molecular Weight (g/mol) 481.5
Formula Weight 481.175