Learn More
Lonza™ PrEGM™ Prostate Epithelial Cell Growth Medium SingleQuots™ Supplements and Growth Factors
Product Code. 11695500
Change view
Click to view available options
Quantity:
BPE: 2.0 mL, Triiodothyronine: 0.5 mL, Insulin: 0.5 mL, hEGF: 0.5 mL, Hydrocortisone: 0.5 mL, Transferrin: 0.5 mL, Epinephrine: 0.5 mL, GA-1000: 0.5 mL, Retinoic Acid: 0.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11695500 BPE: 2.0 mL, Triiodothyronine: 0.5 mL, Insulin: 0.5 mL, hEGF: 0.5 mL, Hydrocortisone: 0.5 mL, Transferrin: 0.5 mL, Epinephrine: 0.5 mL, GA-1000: 0.5 mL, Retinoic Acid: 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11695500 Supplier Lonza™ Supplier No. CC4177
  • ONLINE EXCLUSIVE PRICE
    560.00€
    444.08€ / 1 pièces
    Économisez 115.92€
    20% de réduction
 Restricted Item
Expédition estimée: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Supplements and growth factors kit for use with prostate epithelial cell basal medium.
  • Separately sold SingleQuot™ kit for supplementing basal medium.
  • Part of the full PrEGM™ -2 Prostate Epithelial Cell Growth Medium BulletKit™ (catalog number CC-3166), which consists of basal medium and supplements packaged together.
  • Includes the following:
    • 1 x Orange Cap Vial with BPE 2.00 mL
    • 1 x Amber Vial with Triiodothyronine 0.50 mL
    • 1 x Lilac Cap Vial with Insulin 0.50 mL
    • 1 x Green Cap Vial with hEGF 0.50 mL
    • 1 x Natural Cap Vial with Hydrocortisone 0.50 mL
    • 1 x Natural Cap Vial with Transferrin 0.50 mL
    • 1 x Amber Vial with Epinephrine 0.50 mL
    • 1 x Red Cap Vial with GA-1000 0.50 mL
    • 1 x Amber Vial with Retinoic Acid 0.50 mL
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Epithelial Cells
Product Type Supplements and growth factors
Quantity BPE: 2.0 mL, Triiodothyronine: 0.5 mL, Insulin: 0.5 mL, hEGF: 0.5 mL, Hydrocortisone: 0.5 mL, Transferrin: 0.5 mL, Epinephrine: 0.5 mL, GA-1000: 0.5 mL, Retinoic Acid: 0.5 mL
Content And Storage BPE, Triiodothyronine, Insulin, hEGF, Hydrocortisone, Transferrin, Epinephrine, GA-1000, Retinoic Acid