Learn More
Invitrogen™ protein A, Alexa Fluor™ 546 conjugate
Product Code. 10423632
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10423632 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10423632 Supplier Invitrogen™ Supplier No. P11049
 Restricted Item
Expédition estimée: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
protein A, Alexa Fluor™ 546 conjugate
Our Alexa Fluor 546 protein A conjugate binds to the Fc portion of various immunoglobulins from several different species. This exceptionally bright, orange-fluorescent probe (absorption/emission maxima ∼556/573 nm) can be used to visualize antibodies in a manner analogous to a labeled species-specific secondary antibody.
TRUSTED_SUSTAINABILITY
Specifications
Conjugate Alexa Fluor 546
Quantity 1 mg
Product Line Alexa Fluor
Form Lyophilized Powder
Protein Subtype Protein A
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
For Research Use Only. Not for use in diagnostic procedures.