Learn More
Invitrogen™ protein G, horseradish peroxidase conjugate
Product Code. 10216912
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10216912 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10216912 Supplier Invitrogen™ Supplier No. P21041
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
protein G, horseradish peroxidase conjugate
The horseradish peroxidase (HRP) conjugate of protein G can be used to detect the Fc portion of various classes and subclasses of immunoglobulins from a variety of species. Protein G is a bacterial protein similar to protein A but with superior selectivity for mouse and human IgG.
TRUSTED_SUSTAINABILITY
Specifications
Conjugate HRP
Quantity 1 mg
Form Lyophilized Powder
Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Room Temperature
compliance-icons
Product Identifier
  • protein G, horseradish peroxidase conjugate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: thimerosal
For Research Use Only. Not for use in diagnostic procedures.