Learn More
Bohlender™ PTFE Screw-In Tubing Connectors
Product Code. 10277591
Click to view available options
Thread Size:
G 0.25 in.
NPT 0.25 in.
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 10277591
Brand: Bohlender™ D57904
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
A straight fitting made of PTFE with one connector for elastic tubing and one screw-in thread of either NPT or G.
  • Providing temperature resistance from −200° to +250°C
  • Universal chemical resistance
  • FDA conformity
  • Vacuum suitable
  • For elastic tubing such as Viton™, Tygon™ and silicone
  • Available in different sizes
Specifications
Certifications/Compliance FDA
Description Wrench size 15 mm
Diameter (Metric) Bore 5.0 mm
Material PTFE
Overall Length (Metric) 22 mm
For Use With (Application) Connecting elastic tubing
Min. Operating Temperature (Metric) -200°C
Tubing Size 6.8 mm O.D.
Thread Size NPT 0.25 in.
Shape Straight
Color White
Max. Operating Temperature (Metric) 250°C
Product Type Tubing Connector