Learn More
Fisherbrand™ PTFE Total Immersion Temperature Probe
Product Code. 16226071 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
50 x 5 mm
Length (Metric):
2 m (Cable)
4 m (Cable)
5 m (Cable)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Length (Metric) Unit Size
16226071 50 x 5 mm 5 m (Cable) Each
16206071 50 x 5 mm 2 m (Cable) Each
16216071 50 x 5 mm 4 m (Cable) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16226071
Expédition estimée: 19-01-2027
Add to basket
Add to basket
Fisherbrand™ PTFE Total Immersion Temperature Probe are suitable for use in autoclaves and most corrosive liquids.
  • Sensor - PRT100 Class A
  • Cable - 4 Core PFA insulated with white PFA oversheath
  • Cable fused to body
  • Supplied without plug
  • Inert and non contaminating
  • Pressure Resistant
  • Temperature Range -180°C to +280°C
Specifications
Description Fisherbrand™ PTFE Total Immersion Temperature Probe
Material PTFE
Quantity 1
Sensor Types PRT100 Class A
Temperature Range (Metric) -180°C to +280°C
Dimensions (L x W) 50 x 5 mm
Length (Metric) 5 m (Cable)
Width (Metric) 5 mm