Learn More
BUCHI Pure Chromatography System
Product Code. 17829842 Shop All Buchi Products
Change view
Click to view available options
Type:
Flash/Prep HPLC/UV-Vis/ELSD
Flash/UV
Flash/UV/ELSD
Prep HPLC/UV-Vis
Max. Pressure:
300 bar (4,350 psi)
300 bar (4350 psi)
50 bar (725 psi)
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Max. Pressure Unit Size
17829842 Flash/UV/ELSD 50 bar (725 psi) Each
17527306 Flash/UV 50 bar (725 psi) Each
18568034 Flash/Prep HPLC/UV-Vis/ELSD 300 bar (4,350 psi) Each
17742709 Prep HPLC/UV-Vis 300 bar (4350 psi) Each
17783939 Flash/Prep HPLC/UV-Vis/ELSD 300 bar (4350 psi) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17829842 Supplier BUCHI Supplier No. 11C81500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Extremely compact and can be used outside of the fume hood. Choose from flash systems or combination flash/prep HPLC capable of small to large scale purification of complex mixtures. UV detection is standard or choose to add ELS detection to detect all compounds.
  • Pure space. Pure safety. Pure simplicity.
  • Small footprint onboard solvent platform
  • Cartridge/ column holders on front of the system
  • Lowest sample loss for ELSD (30 uL/min)
  • Closed fraction collector bay
  • Leak and pressure sensors for safety
  • Remote control capable from PC, tablet or smartphone
  • All important parts are easily accessable for cleaning
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Application) Separations
    Height (Metric) 680 mm
    Length (Metric) 570 mm
    Material Stainless Steel
    Max. Pressure 50 bar (725 psi)
    Type Flash/UV/ELSD
    Voltage 100/240 V
    Width (Metric) 365 mm
    Particle Size 40 to 63 μm
    Frequency 50/60 Hz