Learn More
BUCHI Pure Essential Chromatography System
Product Code. 18087401 Shop All Buchi Products
Change view
Click to view available options
Construction:
With Detector
Without Detector
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Construction Unit Size
18087401 With Detector Each
18802722 Without Detector -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 18087401 Supplier BUCHI Supplier No. 11C90010
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
The Pure Essential Chromatography system focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit from increased speed and endless possibilities.
Perform straightforward separations
Many flash separations only require a basic setup. At the bare minimum, a pump for smooth solvent and sample flow, and optionally a UV detector and fraction collector:
  • The Pure Essential Chromatography system benefits from a basic design and setup
  • The small footprint of the instrument allows for placement even in crowded labs, and its robustness makes it highly reliable
  • Basic but powerful and safe thanks to a 50 bar (725 psi) pump, pressure sensors, and regulators
A modular and scalable solution that grows with your needs
  • Every beginning is small. BUCHI supports you in every step of your research journey and offers the highest flexibility for your purification step:
  • Expansion possibilities include the addition of a UV detector and fraction collector to the Pure Chromatography C-900
  • There are no limits regarding the size of the collected fractions
  • Additional options allow you to be flexible regarding sample loading (liquid and solid) and the cartridge holder (any size between 4 to 5000 g)
Easy to use, plug and play chromatography
The Pure Essential Chromatography systems make flash separations easier than ever before; This is due to several key features related to the hardware and software:
  • Intuitive navigation provided by novel and state-of-the-art software (methods, run data, and general settings)
  • All tubing and parts of the instrument are easily accessible and allow the user to follow and monitor the process flow
  • Automation simplifies the entire separation process. It supports safe and efficient operation and reduces any worries about unforeseeable events
TRUSTED_SUSTAINABILITY
Specifications
Construction With Detector
Type Flash/Prep HPLC/UV-Vis/ELSD