Learn More
Karl Hecht™ Pycnometers with Ground-In Thermometer
Product Code. 10504432
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10504432 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10504432 Supplier Karl Hecht™ Supplier No. 42572032
  • ONLINE EXCLUSIVE PRICE
    274.77€
    170.01€ / 1 pièces
    Économisez 104.76€
    38% de réduction
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For the measurement of liquid density and temperature in the laboratory.
  • With ground-in thermometer
  • Precisely adjusted
  • Side capillary with ground-on cap joint size 7.5
  • Flask of Erlenmeyer form
  • Thermometer ground joint size 10/19
  • Thermometer with blue filling, +10°C + 35°C
Specifications
Type Pycnometer
Capacity (Metric) 50 mL
For Use With (Application) Measuring liquid density, temperature
Product Type Blue thermometer filling
Description With ground-in thermometer