Learn More
Pyrrolidine-2-thione, 98%, BLD Pharm™
Product Code. 30945418
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30945418 250 mg 250mg
30932322 25 g -
30929267 5 g 5g
30938030 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30945418 Supplier BLD Pharm Supplier No. BD149933250mg
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 2295-35-4
Molecular Formula C4H7NS
MDL Number MFCD00049351
Quantity 250 mg
SMILES S=C1NCCC1
Molecular Weight (g/mol) 101.17
Percent Purity 98%
Chemical Name or Material Pyrrolidine-2-thione