Learn More
R&D Systems™ Quantikine Immunoassay Control Set1076 for Human VE-Cadherin
Product Code. 30001855
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001855 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001855 Supplier R&D Systems™ Supplier No. QC184
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / 1 pièces
    Économisez 23.48€
    10% de réduction
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For use with catalog number DCADV0
Quantikine Immunoassay Control Set1076 for Human VE-Cadherin is designed for use as quantitative controls for the determination of cytokine concentrations in biological fluids.
  • Applications: ELISA, Control
  • Species Reactivity: Human
  • Form: Lyophilized
  • Composition: Recombinant human cytokines in a buffered protein solution with preservatives
  • Storage: Store unopened product at 2°C to 8°C
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes VE-Cadherin
Sample Type Biological Fluids
Type Immunoassay Control Set
Gene ID (Entrez) 1003
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized