Learn More
Bellingham Stanley™ Quartz Control Plate
Product Code. 11900108
Change view
Click to view available options
Amperage:
5.2 A
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Amperage Unit Size
11900108 5.2 A Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11900108 Supplier Bellingham Stanley™ Supplier No. 3421
Expédition estimée: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High quality quartz control plates for verification of polarimeters
Bellingham and Stanley offer a number of traceable calibration materials for verifying and calibrating refractometers. AG fluids and calibration oils are supplied in packs of 5 x 5mL glass bottles and accompanied by a UKAS Certificate of Calibration showing values at 20°C and 589.3nm, these certified reference materials provide an ideal solution to your refractometer calibration needs. Quartz Control Plates (QCP) are also offered for verifying and calibrating polarimeters and are supplied with a Certificate of Conformity showing values at sodium and NIR for both angular and sugar scales. A thermal block is available for use with Bellingham and Stanley ADP polarimeters, it houses the external temperature sensor enabling the use of quartz temperature compensation. QCPs can be supplied with UKAS Certificate of Calibration showing traceability to PTB as an optional service.
For temperatures other than 20°C, both RI and Brix values are available from the manufacturer's website.
RI and Brix values
  • Wide choice of values
  • 12 month validity
  • UKAS Certificate of Calibration
  • Traceable to NIST and ICUMSA
Thermal block for use with ADP/S available, please enquire.
TRUSTED_SUSTAINABILITY
Specifications
Type Quartz Control Plate
Material Quartz
Amperage 5.2 A